B8252
Bromonucleic acid
Synonym(s):
2-Bromostearic Acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106305
PubChem Substance ID:
extent of labeling
~6% Br (by weight)
storage temp.
−20°C
Application
2-Bromostearate acid (bromonucleic acid) is used to inhibit fatty acid oxidation. 2-Bromostearate acid is a mitochondrial carnitine palmitoyltransferase I (CPT I) inhibitor.
Preparation Note
Bromination of yeast RNA.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C R Bliss et al.
The American journal of physiology, 263(5 Pt 1), E890-E896 (1992-11-01)
The development of glucose-stimulated insulin release and time-dependent potentiation (TDP) has been studied in isolated islets from 7-, 14-, and 21-day-old and 3-mo-old rats. Responses were small at 7 days and changed little at 14 days. At 21 days the
Effect of 2-bromostearate on glucose-phosphorylating activities and the dynamics of insulin secretion in islets of Langerhans during fasting.
Bedoya FJ, Ramirez R, Arilla E, Goberna R.
Diabetes, 33, 858-863 (1084)
J Tamarit-Rodriguez et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 16(3), 115-119 (1984-03-01)
The hypothesis was made of an increased oxidation of fatty acids (FFA) and a decrease of their esterification rate contributing to the islet secretory defect during starvation. 2-Bromostearate (BrS), a FFA-oxidation inhibitor, was therefore tested on the islet secretion of
S Chen et al.
Diabetes, 43(7), 878-883 (1994-07-01)
We sought to explore the emerging concept that malonyl-CoA generation, with concomitant suppression of mitochondrial carnitine palmitoyltransferase I (CPT I), represents an important component of glucose-stimulated insulin secretion (GSIS) by the pancreatic beta-cell (Prentki M, Vischer S, Glennon MC, Regazzi
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service