Skip to Content
Merck
All Photos(2)

Key Documents

B8011

Sigma-Aldrich

DL-Benzylsuccinic acid

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O4
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

powder

SMILES string

OC(=O)CC(Cc1ccccc1)C(O)=O

InChI

1S/C11H12O4/c12-10(13)7-9(11(14)15)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,13)(H,14,15)

InChI key

GTOFKXZQQDSVFH-UHFFFAOYSA-N

Gene Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H R Beller et al.
Applied and environmental microbiology, 58(9), 3192-3195 (1992-09-01)
Two dead-end metabolites of anaerobic toluene transformation, benzylsuccinic acid and benzylfumaric acid, accumulated in sulfate-reducing enrichment cultures that were fed toluene as the sole carbon source. Stable isotope-labeled toluene and gas chromatography-mass spectrometry were used to confirm that the compounds
D R Webster et al.
Journal of cellular biochemistry, 60(3), 424-436 (1996-03-01)
alpha-tubulin subunits within microtubules (MTs) can be post-translationally detyrosinated by a tubulin-specific carboxypeptidase (TCP) activity to form biochemically distinct MTs. Attempts to characterize and purify TCP have suffered from the inability to detect low levels of activity and to distinguish
S Mangani et al.
Journal of molecular biology, 223(2), 573-578 (1992-01-20)
The X-ray crystal structure of the carboxypeptidase A-L-benzylsuccinate complex has been refined at 2.0 A resolution to a final R-factor of 0.166. One molecule of the inhibitor binds to the enzyme active site. The terminal carboxylate forms a salt link
D E Reusser et al.
Journal of chromatography. A, 953(1-2), 215-225 (2002-06-13)
Benzylsuccinic acid (BSA) and methylbenzylsuccinic acid (methyl-BSA) are unambiguous biotransformation products resulting from anaerobic toluene and xylene biodegradation, respectively. A solid-phase extraction method based on polystyrene-divinylbenzene sorbent was developed for the quantitative BSA determination in groundwater samples as an alternative
D M Ferry et al.
Gastroenterology, 97(1), 61-67 (1989-07-01)
Intestinal absorption and enterohepatic circulation of N-formyl-methionyl-leucyl-125I-tyrosine, a bioactive synthetic analog of the bacterial chemotactic peptide N-formyl-methionyl-leucyl-phenylalanine has been investigated in the rat. In ileum and proximal and distal colon, dithiothreitol, which increases mucosal permeability, increased peptide absorption and biliary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service