Skip to Content
Merck
All Photos(1)

Key Documents

B6009

Sigma-Aldrich

Biphenyl-4-carbonyl chloride

Synonym(s):

4-Phenylbenzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4COCl
CAS Number:
Molecular Weight:
216.66
Beilstein:
472842
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

mp

110-112 °C (lit.)

storage temp.

−20°C

SMILES string

ClC(=O)c1ccc(cc1)-c2ccccc2

InChI

1S/C13H9ClO/c14-13(15)12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H

InChI key

JPVUWCPKMYXOKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service