Skip to Content
Merck
All Photos(1)

Key Documents

B5397

Sigma-Aldrich

[Tyr4]-Bombesin

≥97% (HPLC), suitable for ligand binding assays

Synonym(s):

pGlu-Gln-Arg-Tyr-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H108N24O19S
CAS Number:
Molecular Weight:
1669.86
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

[Tyr4]-Bombesin, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)CNC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(N)=O)NC(=O)CNC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]5CCC(=O)N5)C(C)C)C(N)=O

InChI

1S/C74H108N24O19S/c1-36(2)26-50(70(114)91-45(62(78)106)23-25-118-6)95-71(115)53(29-41-32-81-35-86-41)89-60(105)34-85-73(117)61(37(3)4)98-63(107)38(5)87-69(113)52(28-40-31-83-44-11-8-7-10-43(40)44)97-68(112)49(18-21-56(76)101)94-72(116)54(30-57(77)102)90-59(104)33-84-64(108)51(27-39-13-15-42(99)16-14-39)96-65(109)46(12-9-24-82-74(79)80)92-67(111)48(17-20-55(75)100)93-66(110)47-19-22-58(103)88-47/h7-8,10-11,13-16,31-32,35-38,45-54,61,83,99H,9,12,17-30,33-34H2,1-6H3,(H2,75,100)(H2,76,101)(H2,77,102)(H2,78,106)(H,81,86)(H,84,108)(H,85,117)(H,87,113)(H,88,103)(H,89,105)(H,90,104)(H,91,114)(H,92,111)(H,93,110)(H,94,116)(H,95,115)(H,96,109)(H,97,112)(H,98,107)(H4,79,80,82)/t38-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,61-/m0/s1

InChI key

QORXQELQSJVKIL-XLSKZGPMSA-N

Gene Information

human ... GRP(2922)

Amino Acid Sequence

Glp-Gln-Arg-Tyr-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2

Biochem/physiol Actions

[Lys3]-Bombesin and [Tyr4]-Bombesin are Bombesin analogues used to differentiate and study the bombesin receptors BBR1, BBR2 and BBR3 and the homologue receptors, gastrin-releasing peptide (GRP) receptor (GRPR).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Rogers et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 38(8), 1221-1229 (1997-08-01)
The gastrin releasing peptide receptor (GRPr) has a high affinity for the 14 amino acid bombesin peptide. For this analysis, [125I]-Tyr4-bombesin was compared with [125I]-mIP-bombesin (a seven amino acid bombesin analog) for in vitro binding and internalization into tumor cells
C Tang et al.
British journal of cancer, 75(10), 1467-1473 (1997-01-01)
Gut hormones that modulate the growth of normal pancreas may also modulate the growth of cancers originating from pancreas. This study visualized and compared the receptors for cholecystokinin (CCK), bombesin (BBS), secretin and vasoactive intestinal peptide (VIP) in tumour-free tissue
G Halmos et al.
Cancer letters, 85(1), 111-118 (1994-09-30)
Binding of the radiolabeled bombesin analog [125I-Tyr4]bombesin to crude cell membranes of MKN45 human gastric cancer grown in nude mice was investigated in vitro. Scatchard analyses of multipoint binding data, performed by complete displacement method demonstrated the presence of two
S R Preston et al.
British journal of cancer, 71(5), 1087-1089 (1995-05-01)
Human colorectal cancer tissue and matched uninvolved mucosa from 21 patients were examined by radioligand displacement for the presence of binding sites for bombesin-like peptides. Five cancers, but no uninvolved mucosa, expressed high-affinity, low-capacity bombesin binding sites (Kd = 6.53
J L Scemama et al.
Regulatory peptides, 13(2), 125-132 (1986-01-01)
The binding of bombesin to its receptors on normal human pancreatic membranes was investigated using high specific activity, radioiodinated bombesin ([125I]-Tyr4-bombesin), prepared by an oxidative method with chloramine-T. Binding was specific, temperature-dependent, saturable, reversible and linearly related to membranes protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service