Skip to Content
Merck
All Photos(1)

Documents

B4312

Sigma-Aldrich

BTK, active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AGMX1, AT, ATK, BPK, IMD1, PSCTK, XLA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

37-49 nmol/min·mg

mol wt

~78 kDa

solubility

water: soluble

UniProt accession no.

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BTK(695)

Biochem/physiol Actions

BTK (also known as Bruton tyrosine kinase) plays a crucial role in B-lymphocyte differentiation and activation. BTK interacts with SRC homology 3 domains of FYN, LYN and HCK that are activated upon stimulation of B- and T-cell receptors.Defects in the BTK gene cause Agammaglobulinemia, an X-linked immunodeficiency characterized by failure to produce mature B lymphocyte cells and associated with a failure of Ig heavy chain rearrangement. The unique role of BTK makes it a desirable target for potential anti-cancer, anti-inflammatory and anti-viral agents as well as other treatments.

Physical form

Supplied in 50mM NaPhosphate pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1mM PMSF, 0.2 mM DTT, 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A O Vassilev et al.
Current pharmaceutical design, 10(15), 1757-1766 (2004-06-08)
BTK (Bruton's tyrosine kinase) is a member of the TEC family of tyrosine kinases that plays a central but diverse modulatory role in various cellular processes. The unique role of BTK in a multitude of signaling pathways, its function as
G Cheng et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(17), 8152-8155 (1994-08-16)
Bruton's tyrosine kinase (Btk) is a recently described B-cell-specific tyrosine kinase. Mutations in this gene lead to human X chromosome-linked agammaglobulinemia and murine X-linked immunodeficiency. Although genetic evidence strongly suggests that Btk plays a crucial role in B-lymphocyte differentiation and
Federica Giordano et al.
Journal of experimental & clinical cancer research : CR, 38(1), 260-260 (2019-06-16)
Lung cancer is still the main cause of cancer death worldwide despite the availability of targeted therapies and immune-checkpoint inhibitors combined with chemotherapy. Cancer cell heterogeneity and primary or acquired resistance mechanisms cause the elusive behaviour of this cancer and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service