Skip to Content
Merck
All Photos(1)

Key Documents

B3806

Sigma-Aldrich

Bisindolylmaleimide VIII acetate salt

≥93% (H-NMR)

Synonym(s):

Ro 31-7549 acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22N4O2 · C2H4O2
CAS Number:
Molecular Weight:
458.51
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥93% (H-NMR)

solubility

DMSO: soluble
H2O: soluble
methanol: soluble

storage temp.

−20°C

SMILES string

CC(O)=O.Cn1cc(C2=C(C(=O)NC2=O)c3cn(CCCN)c4ccccc34)c5ccccc15

InChI

1S/C24H22N4O2.C2H4O2/c1-27-13-17(15-7-2-4-9-19(15)27)21-22(24(30)26-23(21)29)18-14-28(12-6-11-25)20-10-5-3-8-16(18)20;1-2(3)4/h2-5,7-10,13-14H,6,11-12,25H2,1H3,(H,26,29,30);1H3,(H,3,4)

InChI key

VEOXVBTXROWDAH-UHFFFAOYSA-N

Biochem/physiol Actions

Bisindolylmaleimide VIII acetate is a selective inhibitor of protein kinase C (PKC). Bisindolylmaleimide VIII acetate enhances Fas- and TRAIL-mediated apoptosis and inhibits T cell-mediated autoimmune diseases.
Selective inhibitor of protein kinase C. Enhances Fas- and TRAIL-mediated apoptosis and inhibits T cell-mediated autoimmune diseases.

Caution

Photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service