Skip to Content
Merck
All Photos(1)

Key Documents

B178

Sigma-Aldrich

Butein

solid

Synonym(s):

1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one, 2′,3,4,4′-Tetrahydroxychalcone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

yellow

solubility

DMSO: >50 mg/mL
H2O: insoluble

SMILES string

OC1=CC(O)=C(C(/C=C/C2=CC(O)=C(O)C=C2)=O)C=C1

InChI

1S/C15H12O5/c16-10-3-4-11(14(19)8-10)12(17)5-1-9-2-6-13(18)15(20)7-9/h1-8,16,18-20H/b5-1+

InChI key

AYMYWHCQALZEGT-ORCRQEGFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Butein (2′,3,4,4′-tetrahydroxychalcone) is a chalcone and a flavonoid, that is produced in plants. It is a plant polyphenol and a bioactive constituent, that is extracted from the heartwood of Dalbergia odorifera, Caragana jubata and Rhus verniciflua stokes, and the stem bark of cashews (Semecarpus anacardium).[1]

Biochem/physiol Actions

Butein exhibit several pharmacological activities, such as anti-oxidant and anti-inflammatory activity. It stimulates apoptotic cell death of human cervical cancer cells. It has therapeutic potentials for chronic diseases, including liver tuberculosis, obesity, diabetes and hypertension. Butein can repress migration and invasion of bladder, breast and pancreatic cancer cells.[1]
Inhibits EGFR and Src tyrosine kinase activities; inhibits cAMP-dependent PDE-IV. Induces apoptosis in B16 melanoma cells and HL-60 human leukemia cells.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Caution

Photosensitive

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zafar Rasheed et al.
The Journal of pharmacology and experimental therapeutics, 333(2), 354-363 (2010-02-19)
Activation of mast cells in rheumatoid synovial tissue has often been associated with tumor necrosis factor (TNF)-alpha, interleukin (IL)-6, and IL-8 production and disease pathogenesis by adjacent cell types. Butea monosperma (BM) is a well known medicinal plant in India
Karin Schlangen et al.
Journal of experimental botany, 61(12), 3451-3459 (2010-06-23)
A chalcone 3-hydroxylase (CH3H) cDNA clone was isolated and characterized from Cosmos sulphureus petals accumulating butein (2',3,4,4'-tetrahydroxychalcone) derivatives as yellow flower pigments. The recombinant protein catalyses the introduction of an additional hydroxyl group in the B-ring of chalcones, a reaction
S M Yu et al.
European journal of pharmacology, 280(1), 69-77 (1995-06-23)
Butein, isolated from Dalbergia odorifera T. Chen, caused endothelium-dependent relaxation of rat aorta precontracted with phenylephrine. This effect was abolished in endothelium-denuded aorta and in endothelium-intact aorta in the presence of NG-monomethyl-L-arginine, oxyhemoglobin and methylene blue, whereas the effect was
Naghma Khan et al.
Antioxidants & redox signaling, 16(11), 1195-1204 (2011-11-26)
Prostate cancer (PCa) is one of the most common cancers in men in the United States with similar trends worldwide. For several reasons, it is an ideal candidate disease for intervention with dietary botanical antioxidants. Indeed, many botanical antioxidants are
Dong-Oh Moon et al.
Molecular cancer therapeutics, 9(6), 1583-1595 (2010-06-03)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) induces cell death in various types of cancer cells but has little or no effect on normal cells. Human hepatoma cells are resistant to TRAIL-induced apoptosis. Although butein is known to mediate anticancer, anti-inflammatory

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service