Skip to Content
Merck
All Photos(2)

Key Documents

B173

Sigma-Aldrich

BRL 54443 maleate salt

solid

Synonym(s):

3-(1-Methylpiperidin-4-yl)-1H-indol-5-ol maleate salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€162.00

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€162.00

About This Item

Empirical Formula (Hill Notation):
C14H18N2O · C4H4O4
CAS Number:
Molecular Weight:
346.38
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

Quality Level

color

white

solubility

H2O: 50 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.CN1CCC(CC1)c2c[nH]c3ccc(O)cc23

InChI

1S/C14H18N2O.C4H4O4/c1-16-6-4-10(5-7-16)13-9-15-14-3-2-11(17)8-12(13)14;5-3(6)1-2-4(7)8/h2-3,8-10,15,17H,4-7H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

INGCLXPSKXSYND-BTJKTKAUSA-N

Gene Information

Biochem/physiol Actions

Potent 5-HT1E/1F serotonin receptor agonist.

Caution

Photosensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of BRL 54443 (3-(1-methylpiperidin-4-yl)-1H-indol-5-ol) a 5-HT1E/1F receptor agonist on general behaviour and maximal electroshock seizure threshold in the rat.
Lightowler, et al.
British Journal of Pharmacology, 123, 237P-237P (1998)
BRL 54443 a potent agonist with selectivity for human cloned 5-HT1E and 5-HT1F receptors.
Brown, et al.
British Journal of Pharmacology, 123, 233P-233P (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service