Skip to Content
Merck
All Photos(1)

Key Documents

B138

Sigma-Aldrich

BTCP hydrochloride

Synonym(s):

Benocyclidine hydrochloride, N-[1-(Benzo[b]thien-2-yl-cyclohexyl)]piperidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25NS · HCl
CAS Number:
Molecular Weight:
335.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

solubility

H2O: >180 mg/mL
ethanol: soluble

storage temp.

2-8°C

SMILES string

Cl[H].C1CCN(CC1)C2(CCCCC2)c3cc4ccccc4s3

InChI

1S/C19H25NS.ClH/c1-5-11-19(12-6-1,20-13-7-2-8-14-20)18-15-16-9-3-4-10-17(16)21-18;/h3-4,9-10,15H,1-2,5-8,11-14H2;1H

InChI key

XBOZTWUGBRZVBP-UHFFFAOYSA-N

Gene Information

human ... SLC6A3(6531)

Biochem/physiol Actions

BTCP is a potent and selective blocker of dopamine transport with little affinity for phencyclidine sites.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Maurice et al.
Journal of neurochemistry, 56(2), 553-559 (1991-02-01)
The phencyclidine (PCP) derivative, [3H]N-[1-(2-benzo[b]thiophenyl)cyclohexyl]piperidine ([3H]BTCP), was used to label in vivo the dopamine uptake complex in mouse brain. The striatum accumulated the highest level of total and specific binding. Drugs which bind to the dopamine uptake site inhibited [3H]BTCP
C Deleuze-Masquefa et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 9(4), 345-354 (2000-02-09)
1-[1-(2-Benzo[b]thiopheneyl)cyclohexyl]piperidine hydrochloride (BTCP) and cocaine bind to the neuronal dopamine transporter (DAT) to strongly inhibit dopamine (DA) reuptake. Although similar to acute administration, cocaine and BTCP produce sensitization and tolerance, respectively, on chronic administration. We previously found that liver microsomes
G Attali et al.
Brain research, 756(1-2), 153-159 (1997-05-09)
The present study was designed to investigate the modulatory effect of gonadal steroids on brain dopamine (DA) and serotonin (5-HT) presynaptic transporters in female and male rats. Female and male rats were castrated and treated with either vehicle or gonadal
W Koek et al.
The Journal of pharmacology and experimental therapeutics, 250(3), 1019-1027 (1989-09-01)
Phencyclidine (PCP) inhibits dopamine (DA) uptake and acts as a noncompetitive N-methyl-D-aspartate antagonist by binding to PCP receptors. The PCP analog N-[1-(2-benzo(b)thiophenyl) cyclohexyl]piperidine (BTCP, GK13) is a potent DA uptake inhibitor, but has low affinity for PCP receptors. The behavioral
M Froimowitz et al.
Journal of computer-aided molecular design, 14(2), 135-146 (2000-03-18)
Molecular mechanics calculations using MM3-92 and ab initio quantum mechanical calculations using SPARTAN 5.0 were performed on the structurally similar PCP and BTCP, in which only the latter has a cocaine-like pharmacological profile as a dopamine reuptake blocker. Calculations were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service