Key Documents
AXYVT38431ULRS
Pipette tips in 384-tip format, robotic
size 30 μL, pkg of 5x3840tips/cs, racked, sterile
Synonym(s):
Pipette tips for Velocity
Select a Size
1 PKG
€5,130.00
€5,130.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
€5,130.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
size:
30 μL
sterility:
sterile
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc VT-384-31UL-R-S
Corning
€5,130.00
Please contact Customer Service for Availability
sterility
sterile
manufacturer/tradename
Axygen Inc VT-384-31UL-R-S
Corning
packaging
pkg of 5x3840tips/cs
size
30 μL
General description
  • Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
  • Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
  • The clear polypropylene high-quality tips are ideal for sensitive biological applications.
  • RNase-/DNase-free and nonpyrogenic.
Other Notes
Browse all pipette tips
Legal Information
Agilent is a registered trademark of Agilent Technologies, Inc.
Axygen is a registered trademark of Corning, Inc.
Velocity11 VPrep is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)