Skip to Content
Merck
All Photos(1)

Key Documents

AXYTXL10RS

Pipette tips, for Pipetman® P2/P10 and Eppendorf® Research/Reference pipettors, ultramicro, extra long

size 0.1-10 μL, pkg of 5x960tips/cs, racked, clear, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€363.00

€363.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€363.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
0.1—10 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc TXL-10-R-S
Corning

€363.00


Please contact Customer Service for Availability

material

clear

sterility

sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc TXL-10-R-S
Corning

packaging

pkg of 5x960tips/cs

size

0.1-10 μL

volume

0.1—10 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Pipetman is a registered trademark of Gilson S.A.S. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service