Skip to Content
Merck
All Photos(1)

Key Documents

AXYTT200CBKHTRS

Tecan®-style liquid level sensing tips

size 200 μL, case of 2304 ea 24x96tips/cs, racked, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€389.00

€389.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€389.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
200 μL
sterility:
sterile
feature:
DNA free
RNase free
endotoxin free
filter: no
packaging:
case of 2304 ea 24x96tips/cs
manufacturer/tradename:
Corning TT-200-CBK-HTR-S

€389.00


Please contact Customer Service for Availability

material

black

sterility

sterile

feature

DNA free
RNase free
endotoxin free
filter: no

packaging

case of 2304 ea 24x96tips/cs

manufacturer/tradename

Corning TT-200-CBK-HTR-S

size

200 μL

volume

200 μL

Looking for similar products? Visit Product Comparison Guide

Legal Information

Tecan is a registered trademark of Tecan Group AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service