Skip to Content
Merck
All Photos(1)

Key Documents

AXYTT1000CBKHTR24

Tecan®-style liquid level sensing tips

size 1000 μL, pkg of 24x192tips/cs, 2 racks in cardboard box

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€572.00

€572.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€572.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1000 μL
sterility:
non-sterile
feature:
DNA free
RNase free
endotoxin free
filter: no
packaging:
pkg of 24x192tips/cs
manufacturer/tradename:
Corning TT-1000-CBK-HTR-24

€572.00


Please contact Customer Service for Availability

material

black

sterility

non-sterile

feature

DNA free
RNase free
endotoxin free
filter: no

manufacturer/tradename

Corning TT-1000-CBK-HTR-24

packaging

pkg of 24x192tips/cs

size

1000 μL

volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Corning is a registered trademark of Corning, Inc.
Tecan is a registered trademark of Tecan Group AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service