AXYT200CSTK
Pipette tips
stacked racks, size 200 μL, clear, pkg of 10x480tips/cs
Synonym(s):
pipet tips
Select a Size
€289.00
Select a Size
About This Item
Corning
€289.00
Recommended Products
manufacturer/tradename
Axygen Inc T-200-C-STK
Corning
packaging
pkg of 10x480tips/cs
size
200 μL
volume
200 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service