Skip to Content
Merck
All Photos(1)

Key Documents

AXYRRP200CBKRS

Pipette tips for Packard® MPII and Janus®

conductive, racked, capacity 200 μL, black, pkg of 5x960tips/cs, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

capacity:
200 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc RRP-200-CBK-R-S
Corning

sterility

sterile

manufacturer/tradename

Axygen Inc RRP-200-CBK-R-S
Corning

packaging

pkg of 5x960tips/cs

capacity

200 μL

color

black

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Janus is a registered trademark of PerkinElmer Inc.
Packard is a registered trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service