Skip to Content
Merck
All Photos(1)

Key Documents

AXYQT1100RS

Pipette tips for Qiagen®

capacity 1100 μL, clear, racked, pkg of 16x96tips/cs, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1100 μL
sterility:
sterile
packaging:
pkg of 16x96tips/cs
manufacturer/tradename:
Axygen Inc QT-1100-R-S
Corning

sterility

sterile

manufacturer/tradename

Axygen Inc QT-1100-R-S
Corning

packaging

pkg of 16x96tips/cs

capacity

1100 μL

volume

1100 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Qiagen is a registered trademark of Qiagen Group

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service