Key Documents
AXYPCR96FSW
PCR microplate, full skirt
size 96 wells, conical bottom white polypropylene wells, pkg of 5x10plates/cs
Synonym(s):
96 multiwell plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom white polypropylene wells
maximum volume:
200 μL
size:
96 wells
working volume:
200 μL
Pricing and availability is not currently available.
material
conical bottom white polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-96-FS-W
Corning
packaging
pkg of 5x10plates/cs
maximum volume
200 μL
size
96 wells
working volume
200 μL
binding type
non-treated surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)