AXYFXF180RS
Pipette filter tips for Beckman® FX Robotics System
racked, capacity 165 μL, pkg of 5x960tips/cs, clear, sterile
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
Select a Size
1 PKG
€766.00
€766.00
Please contact Customer Service for Availability
All Photos(1)
Select a Size
Change View
1 PKG
€766.00
About This Item
UNSPSC Code:
41121601
NACRES:
NB.02
volume:
165 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc FXF-180-R-S
Corning
Corning
€766.00
Please contact Customer Service for Availability
sterility
sterile
manufacturer/tradename
Axygen Inc FXF-180-R-S
Corning
packaging
pkg of 5x960tips/cs
capacity
165 μL
volume
165 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Legal Information
Beckman is a registered trademark of Beckman Coulter, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service