Skip to Content
Merck
All Photos(1)

Key Documents

AXYFXF180LRS

MAXYMum Recovery® pipette filter tips for Beckman® FX Robotics System

racked, capacity 165 μL, pkg of 5x960tips/cs, clear, sterile

Synonym(s):

Pipette tips for Velocity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
165 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc FXF-180-L-R-S
Corning
Product AXYFXF180LRS is not currently sold in your country. Contact Technical Service

material

clear

sterility

sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter

manufacturer/tradename

Axygen Inc FXF-180-L-R-S
Corning

packaging

pkg of 5x960tips/cs

capacity

165 μL

volume

165 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Clear polypropylene with aerosol-resistant filter tips
  • To ensure consistency in performance and quality, tips are produced to exact standards and are assembled automatically
  • RNase/DNase-free and nonpyrogenic tips
  • Compatible with Biomek® AP-96 head, Beckman Coulter, SPAN-8, MP200, MP20, and AP-384 head

Other Notes

Browse all pipette tips

Legal Information

Beckman is a registered trademark of Beckman Coulter, Inc.
Biomek is a registered trademark of Beckman Instruments, Inc.
Corning is a registered trademark of Corning, Inc.
Coulter is a trademark of Beckman Coulter, Inc.
Maxymum Recovery is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service