Skip to Content
Merck
All Photos(1)

Key Documents

AXYFX384RS

Pipette tips for Beckman® FX Robotics System

capacity 30 μL, pkg of 5x3840tips/cs, clear, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€4,850.00

€4,850.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€4,850.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
30 μL
sterility:
sterile
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc FX-384-R-S
Corning

€4,850.00


Please contact Customer Service for Availability

sterility

sterile

manufacturer/tradename

Axygen Inc FX-384-R-S
Corning

packaging

pkg of 5x3840tips/cs

capacity

30 μL

volume

30 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Beckman is a registered trademark of Beckman Coulter, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service