Skip to Content
Merck
All Photos(1)

Key Documents

AXYFX384LR

MAXYMum Recovery® pipette tips for Beckman® FX Robotics System

racked, capacity 30 μL, pkg of 5x3840tips/cs, clear

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
capacity:
30 μL
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc FX-384-L-R
Corning

manufacturer/tradename

Axygen Inc FX-384-L-R
Corning

packaging

pkg of 5x3840tips/cs

capacity

30 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Beckman is a registered trademark of Beckman Coulter, Inc.
Maxymum Recovery is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service