Skip to Content
Merck
All Photos(1)

Key Documents

AV51676

Sigma-Aldrich

Anti-NUFIP2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-182-FIP, Anti-82-FIP, Anti-FIP-82, Anti-FLJ10976, Anti-KIAA1321, Anti-MGC117262, Anti-Nuclear fragile X mental retardation protein interacting protein 2, Anti-PIG1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

76 kDa

species reactivity

human, dog, rat, bovine, mouse, horse, guinea pig, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUFIP2(57532)

Immunogen

Synthetic peptide directed towards the N terminal region of human NUFIP2

Application

Anti-NUFIP2 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

Nuclear fragile X mental retardation protein interacting protein 2 (NUFIP2) is an RNA-binding protein involved in posttranslational regulation. It is found in association with actively translating polyribosomes, RNA granules in cytoplasm, RNA complexes in neurites and RNAi machinery.

Sequence

Synthetic peptide located within the following region: IPNGVVTNNSGYITNGYMGKGADNDGSGSESGYTTPKKRKARRNSAKGCE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingyue Jia et al.
The Journal of cell biology, 221(11) (2022-10-01)
We report that lysosomal damage is a hitherto unknown inducer of stress granule (SG) formation and that the process termed membrane atg8ylation coordinates SG formation with mTOR inactivation during lysosomal stress. SGs were induced by lysosome-damaging agents including SARS-CoV-2ORF3a, Mycobacterium
Barbara Bardoni et al.
Human molecular genetics, 12(14), 1689-1698 (2003-07-03)
FMRP is an RNA binding protein whose absence produces pathological manifestations of the fragile-X syndrome. FMRP is a component of mRNP complexes found in association with actively translating polyribosomes, RNA complexes trafficking in neurites, RNA granules in cytoplasm and, in
Andres Ramos et al.
Structure (London, England : 1993), 14(1), 21-31 (2006-01-13)
FMRP, whose lack of expression causes the X-linked fragile X syndrome, is a modular RNA binding protein thought to be involved in posttranslational regulation. We have solved the structure in solution of the N-terminal domain of FMRP (NDF), a functionally

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service