Skip to Content
Merck
All Photos(1)

Key Documents

AV51642

Sigma-Aldrich

Anti-PAXIP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CAGF28, Anti-CAGF29, Anti-FLJ41049, Anti-PACIP1, Anti-PAX interacting (with transcription-activation domain) protein 1, Anti-PAXIP1L, Anti-PTIP, Anti-TNRC2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€374.00

€374.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€374.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

74 kDa

species reactivity

rat, guinea pig, rabbit, horse, human, dog, bovine, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAXIP1(22976)

Immunogen

Synthetic peptide directed towards the N terminal region of human PAXIP1

Application

Anti-PAXIP1 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

PAX interacting (with transcription-activation domain) protein 1 (PAXIP1) is a nuclear protein that belongs to the paired box gene family. It is characterized by six breast cancer carboxy-terminal (BRCT) domains. PAXIP1 is involved in chromatin condensation during mitosis and maintenance of genome stability.

Sequence

Synthetic peptide located within the following region: MFDDSSDSSPEKQERNLNWTPAEVPQLAAAKRRLPQGKEPGLINLCANVP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Ah Cho et al.
Molecular and cellular biology, 23(5), 1666-1673 (2003-02-18)
The Pax transactivation domain-interacting protein (PTIP) is a large nuclear protein with multiple BRCT domains that was identified on the basis of its interaction with transcription factors of the Pax and Smad families. To address the function of PTIP during
Ivan M Munoz et al.
Nucleic acids research, 35(16), 5312-5322 (2007-08-11)
Human (h)PTIP plays important but poorly understood roles in cellular responses to DNA damage. hPTIP interacts with 53BP1 tumour suppressor but only when 53BP1 is phosphorylated by ATM after DNA damage although the mechanism(s) and significance of the interaction of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service