Skip to Content
Merck
All Photos(1)

Key Documents

AV49753

Sigma-Aldrich

Anti-FNDC3B antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DKFZp686D14170, Anti-DKFZp762K137, Anti-FAD104, Anti-FLJ23399, Anti-Fibronectin type III domain containing 3B, Anti-MGC10002, Anti-PRO4979, Anti-YVTM2421

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€382.00

€382.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€382.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€382.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

rabbit, horse, rat, dog, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FNDC3B(64778)

Related Categories

Immunogen

Synthetic peptide directed towards the N terminal region of human FNDC3B

Application

Anti-FNDC3B antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

Fibronectin type III domain containing 3B (FNDC3B; FAD104) is a positive regulator of adipogenesis and is highly expressed in the early stage of adipogenesis. FNDC3B gene is reportedly associated with primary open-angle glaucoma.

Sequence

Synthetic peptide located within the following region: RARSSPKSNDSDLQEYELEVKRVQDILSGIEKPQVSNIQARAVVLSWAPP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kei Tominaga et al.
FEBS letters, 577(1-2), 49-54 (2004-11-06)
A novel gene named fad104 (factor for adipocyte differentiation-104), whose expression level quickly increased in the early stage of adipogenesis, was isolated and characterized. The deduced amino acid sequence of fad104 revealed the possible presence of a fibronectin type III
Yi Lu et al.
Nature genetics, 45(2), 155-163 (2013-01-08)
Central corneal thickness (CCT) is associated with eye conditions including keratoconus and glaucoma. We performed a meta-analysis on >20,000 individuals in European and Asian populations that identified 16 new loci associated with CCT at genome-wide significance (P < 5 ×

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service