Skip to Content
Merck
All Photos(1)

Key Documents

AV48996

Sigma-Aldrich

Anti-GPT2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ALT2, Anti-Glutamic pyruvate transaminase (alanine aminotransferase) 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€454.00

€454.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€454.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€454.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58 kDa

species reactivity

human, rat, rabbit, dog, bovine, horse, mouse, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPT2(84706)

Related Categories

Immunogen

Synthetic peptide directed towards the C terminal region of human GPT2

Application

Anti-GPT2 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Glutamic pyruvate transaminase (alanine aminotransferase) 2 (GPT2; ALT2) is an enzyme that participates in the reversible transamination reaction that yields glutamate and pyruvate during gluconeogenesis.

Sequence

Synthetic peptide located within the following region: PEYSSNVELASFHSTSKGYMGECGYRGGYMEVINLHPEIKGQLVKLLSVR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Björn Glinghammar et al.
International journal of molecular medicine, 23(5), 621-631 (2009-04-11)
Serum alanine aminotransferase (ALT) is used as a clinical marker of hepatotoxicity. Three forms of human ALT have been identified, ALT1 and 2 and an alternative splice variant of ALT2 (herein called ALT2_2). The standard ALT activity assay does not
Claudia Billing et al.
Proteomics, 17(15-16) (2017-07-01)
Hematopoietic bone marrow is a regenerative tissue of high clinical relevance, yet relatively little is known about the metabolism of the stem and progenitor populations concerned. We have used a multipotent murine cell line to generate sufficient numbers of cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service