Skip to Content
Merck
All Photos(1)

Key Documents

AV48819

Sigma-Aldrich

Anti-TRMU antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC99627, Anti-MTO2, Anti-MTU1, Anti-TRMT, Anti-TRMT1, Anti-TRNT1, Anti-tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€569.00

€569.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€569.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€569.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

mouse, human, rat, bovine, guinea pig, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRMU(55687)

General description

TRMU codes for tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase that catalyzes the 2-thiolation of uridine. Studies have reported that TRMU mutations affect the deafness-associated mutations in 12S ribosomal RNA.
Rabbit Anti-TRMU antibody recognizes bovine, canine, human, mouse, and rat TRMU.

Immunogen

Synthetic peptide directed towards the C terminal region of human TRMU

Application

Rabbit Anti-TRMU antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

TRMU is a member of the trmU family. It is a mitochondria-specific tRNA-modifying enzyme that is required for the 2-thio modification of 5-taurinomethyl-2-thiouridine tRNA-Lys on the wobble position of the anticodon.This gene is a member of the trmU family. It encodes a mitochondria-specific tRNA-modifying enzyme that is required for the 2-thio modification of 5-taurinomethyl-2-thiouridine tRNA-Lys on the wobble position of the anticodon.

Sequence

Synthetic peptide located within the following region: ALATGQFAVFYKGDECLGSGKILRLGPSAYTLQKGQRRAGMATESPSDSP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min-Xin Guan et al.
American journal of human genetics, 79(2), 291-302 (2006-07-11)
The human mitochondrial 12S ribosomal RNA (rRNA) A1555G mutation has been associated with aminoglycoside-induced and nonsyndromic deafness in many families worldwide. Our previous investigation revealed that the A1555G mutation is a primary factor underlying the development of deafness but is
Qingfeng Yan et al.
Biochemical and biophysical research communications, 342(4), 1130-1136 (2006-03-04)
Nuclear modifier genes have been proposed to modulate the phenotypic manifestation of human mitochondrial 12S rRNA A1491G mutation associated with deafness in many families world-wide. Here we identified and characterized the putative nuclear modifier gene TRMU encoding a highly conserved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service