Skip to Content
Merck
All Photos(2)

Key Documents

AV48320

Sigma-Aldrich

Anti-RSU1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLJ31034, Anti-RSP-1, Anti-Ras suppressor protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€287.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

31 kDa

species reactivity

horse, guinea pig, mouse, dog, human, rat, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RSU1(6251)

General description

RSU1 codes for a Ras suppressor protein that associates with LIM5 domain of PINCH1 and contributes to adhesion-related functions in cells. It regulates p38 signaling and affects cell survival[1].
Rabbit Anti-RSU1 antibody recognizes chicken, bovine, human, mouse, rat, zebrafish, and canine RSU1.

Immunogen

Synthetic peptide directed towards the C terminal region of human RSU1

Application

Rabbit Anti-RSU1 antibody is suitable for western blot applications at a concentration of 1.25 μg/ml and for IHC at 4-8 μg/ml.

Biochem/physiol Actions

RSU1 is a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the protein was initially isolated based on its ability to inhibit v-Ras transformation.This gene encodes a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the encoded protein was initially isolated based on its ability to inhibit v-Ras transformation. Multiple alternatively spliced transcript variants for this gene have been reported; one of these variants was found only in glioma tumors.

Sequence

Synthetic peptide located within the following region: PIADQFQLGVSHVFEYIRSETYKYLYGRHMQANPEPPKKNNDKSKKISRK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reyda Gonzalez-Nieves et al.
Journal of cell communication and signaling, 7(4), 279-293 (2013-06-15)
Cell adhesion and migration are complex processes that require integrin activation, the formation and dissolution of focal adhesion (FAs), and linkage of actin cytoskeleton to the FAs. The IPP (ILK, PINCH, Parvin) complex regulates FA formation via binding of the
Gerard W Dougherty et al.
Experimental cell research, 306(1), 168-179 (2005-05-10)
Rsu-1 is a highly conserved leucine rich repeat (LRR) protein that is expressed ubiquitously in mammalian cells. Rsu-1 was identified based on its ability to inhibit transformation by Ras, and previous studies demonstrated that ectopic expression of Rsu-1 inhibited anchorage-independent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service