Skip to Content
Merck
All Photos(1)

Documents

AV48046

Sigma-Aldrich

Anti-YWHAZ antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-KCIP-1, Anti-MGC111427, Anti-MGC126532, Anti-MGC138156, Anti-Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

28 kDa

species reactivity

rat, mouse, horse, rabbit, human, guinea pig, dog, bovine, sheep

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YWHAZ(7534)

General description

Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta (YWHAZ) is a member of the 14-3-3 family that regulates signal transduction. Increased expression of YWHAZ has been linked to the growth and metastasis of gastric carcinoma. YWHAZ amplification is associated with breast cancer recurrence and chemotherapy resistance.
Rabbit Anti-YWHAZ antibody recognizes chicken, bovine, pig, human, mouse, rat, and canine YWHAZ.

Immunogen

Synthetic peptide directed towards the C terminal region of human YWHAZ

Application

Rabbit Anti-YWHAZ antibody is suitable for western blot applications at a concentration of 5μg/ml.

Biochem/physiol Actions

YWHAZ belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and sheep orthologs. The protein interacts with IRS1 protein, suggesting a role in regulating insulin sensitivity.This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and sheep orthologs. The encoded protein interacts with IRS1 protein, suggesting a role in regulating insulin sensitivity. Two transcript variants differing in the 5′ UTR, but encoding the same protein, have been identified for this gene.

Sequence

Synthetic peptide located within the following region: AFDEAIAELDTLSEESYKDSTLIMQLLRDNLTLWTSDTQGDEAEAGEGGE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Li et al.
Nature medicine, 16(2), 214-218 (2010-01-26)
Adjuvant chemotherapy for breast cancer after surgery has effectively lowered metastatic recurrence rates. However, a considerable proportion of women suffer recurrent cancer at distant metastatic sites despite adjuvant treatment. Identification of the genes crucial for tumor response to specific chemotherapy
Y Nishimura et al.
British journal of cancer, 108(6), 1324-1331 (2013-02-21)
Several studies have demonstrated that YWHAZ (14-3-3ζ), included in the 14-3-3 family of proteins, has been implicated in the initiation and progression of cancers. We tested whether YWHAZ acted as a cancer-promoting gene through its activation/overexpression in gastric cancer (GC).

Articles

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service