Skip to Content
Merck
All Photos(2)

Documents

AV46773

Sigma-Aldrich

Anti-SC4MOL antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DESP4, Anti-ERG25, Anti-MGC104344, Anti-Sterol-C4-methyloxidase-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

pig, rabbit, dog, human, horse, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SC4MOL(6307)

General description

Sterol-C4-methyloxidase-like/methylsterol monooxygenase 1 (SC4MOL, DESP4, ERG25, MSMO1) is an endoplasmic reticulum enzyme that catalyzes the demethylation of C4-methlysterols (meiosis-activating sterols, MAS) in the cholesterol synthesis pathway. Defective/mutated SC4MOL contributes to psoriasiform dermatitis, microcephaly, and developmental delay.

Specificity

Anti-SC4MOL polyclonal antibody reacts with zebrafish, bovine, pig, human, mouse, rat, and canine sterol-C4-methyloxidase-like proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human SC4MOL

Application

Anti-SC4MOL polyclonal antibody is used to tag sterol-C4-methyloxidase-like for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of sterol-C4-methyloxidase-like in the management of C4-methlysterols (meiosis-activating sterols, MAS) levels.

Biochem/physiol Actions

Sterol-C4-mehtyl oxidase-like protein was isolated based on its similarity to the yeast ERG25 protein. It contains a set of putative metal binding motifs with similarity to that seen in a family of membrane desaturases-hydroxylases.Sterol-C4-mehtyl oxidase-like protein was isolated based on its similarity to the yeast ERG25 protein. It contains a set of putative metal binding motifs with similarity to that seen in a family of membrane desaturases-hydroxylases. The protein is localized to the endoplasmic reticulum membrane and is believed to function in cholesterol biosynthesis. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: MATNESVSIFSSASLAVEYVDSLLPENPLQEPFKNAWNYMLNNYTKFQIA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service