Skip to Content
Merck
All Photos(3)

Key Documents

AV41643

Sigma-Aldrich

Anti-AGER (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Advanced glycosylation end product-specific receptor, Anti-MGC22357, Anti-RAGE

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€325.00

€325.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€325.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€325.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

glycosylation

Gene Information

human ... AGER(177)

General description

The receptor for advanced glycation end products (RAGE or AGER), a member of the immunoglobulin superfamily, is involved in pathologies such as atherosclerosis, micro- and macrovascular diseases, inflammatory diseases and diabetes. RAGE binds advanced glycation end products (AGE) produced by non-enzymatic glycation or glyoxidation of proteins, lipids and nucleic acids.

The previously assigned protein identifier A2BFI7 has been merged into Q15109. Full details can be found on the UniProt database.

Specificity

Anti-RAGE (AB2) polyclonal antibody reacts with human advanced glycosylation end product-specific receptor (AGER).

Immunogen

Synthetic peptide directed towards the C terminal region of human AGER

Application

Anti-AGER (AB2) polyclonal antibody is used to tag advanced glycation end product receptor protein(s) for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. Anti-AGER (AB2) polyclonal antibody may be used to study the effects/roles of glycation and glyoxidation in cardiovascular disease and diabetes.

Biochem/physiol Actions

AGER mediates interactions of advanced glycosylation end products (AGE). These are nonenzymatically glycosylated proteins which accumulate in vascular tissue in aging and at an accelerated rate in diabetes. It is receptor for amyloid beta peptide.This gene encodes a member of the immunoglobulin superfamily of cell surface molecules. It is a receptor for various molecules, including the amyloidogenic form of serum amyloid A, amyloid-beta protein, members of the S100/calgranulin superfamily and advanced glycation end products. The gene lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Alternative splicing results in two transcript variants encoding different isoforms.

Sequence

Synthetic peptide located within the following region: RIRAGNSSPGPGDPGRPGDSRPAHWGHLVAKAATPRRGEEGPRKPGGRGG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service