Skip to Content
Merck
All Photos(4)

Documents

AV41388

Sigma-Aldrich

Anti-SLC12A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BSC1, Anti-MGC48843, Anti-NKCC2, Anti-Solute carrier family 12 (sodium/potassium/chloride transporters), member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47 kDa

species reactivity

rabbit, rat, human, mouse, pig, dog, bovine, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC12A1(6557)

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC12A1

Biochem/physiol Actions

The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henle′s loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene.

Sequence

Synthetic peptide located within the following region: NEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Fickert et al.
Hepatology (Baltimore, Md.), 58(6), 2056-2069 (2013-07-03)
Tubular epithelial injury represents an underestimated but important cause of renal dysfunction in patients with cholestasis and advanced liver disease, but the underlying mechanisms are unclear. To address the hypothesis that accumulation and excessive alternative urinary elimination of potentially toxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service