Skip to Content
Merck
All Photos(3)

Key Documents

AV40721

Sigma-Aldrich

Anti-HNRPH3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Heterogeneous Nuclear Ribonucleoprotein H3 (2H9)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€287.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

guinea pig, bovine, rabbit, rat, human, horse, mouse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNRPH3(3189)

General description

Heterogeneous Nuclear Ribonucleoproteins (hnRNP) are RNA binding proteins that form complexes with heterogeneous nuclear RNA (hnRNA). HnRNPs regulate pre-mRNA processing, metabolism and nuclear cytoplasmic shuttling. Specific HnRNPs have unique nucleic acid binding properties. Heterogeneous Nuclear Ribonucleoprotein H3 has been identified as an autoantigen for acute anterior uveitis. Heterogeneous Nuclear Ribonucleoprotein H3 may be involved in pre-mRNA splicing associated with inflammatory bowel disease (IBD).

Specificity

Anti-HNRPH3 polyclonal antibody reacts with canine, chicken, zebrafish, human, mouse, rat, and bovine Heterogeneous Nuclear Ribonucleoprotein H3 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human HNRPH3

Application

Anti-HNRPH3 polyclonal antibody is used to tag the heterogeneous nuclear ribonucleoprotein H3 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of heterogeneous nuclear ribonucleoprotein 3 in diseases such as acute anterior uveitis and inflammatory bowel disease (IBD).

Biochem/physiol Actions

HNRPH3 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two repeats of quasi-RRM domains that bind to RNAs. It is localized in nuclear bodies of the nucleus. This protein is involved in the splicing process and it also participates in early heat shock-induced splicing arrest by transiently leaving the hnRNP complexes.This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. It is localized in nuclear bodies of the nucleus. This protein is involved in the splicing process and it also participates in early heat shock-induced splicing arrest by transiently leaving the hnRNP complexes. Multiple alternative transcript variants seem to be present for this gene and some appear to have intronic regions in the mRNA. Presently, only two transcript variants are fully described.

Sequence

Synthetic peptide located within the following region: DYQGRSTGEAFVQFASKEIAENALGKHKERIGHRYIEIFRSSRSEIKGFY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service