Skip to Content
Merck
All Photos(2)

Key Documents

AV40136

Sigma-Aldrich

Anti-EHF antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-9030625L19Rik, Anti-AU019492, Anti-Ets homologous factor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

rat, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ehf(13661)

General description

ETS transcription factors share a conserved DNA-binding "ETS" domain and include several oncoproteins that induce tumorigenesis when overexpressed. ETS homologous factor (EHF) has been found in kidney, lung and somatotroph tumors. ETS, potentially a tumor suppressor and senescence modulator, is most highly expressed in the organs known to form carcinomas upon 11p12 deletion. ETS may be a prognostic marker of carcinomas such as ovarian carcinoma.

Specificity

Anti-EHF polyclonal antibody reacts with human, mouse, rat, chicken, bovine, and canine ETS homologous factor proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of mouse Ehf

Application

Anti-EHF polyclonal antibody is used to tag ETS homologous factor proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of ETS homologous factor as a potential prognostic marker for carcinomas such as ovarian carcinoma cancer and to study gene regulation during carcinogenesis.

Biochem/physiol Actions

Ehf is a transcriptional activator that may play a role in regulating epithelial cell differentiation and proliferation. It may act as a repressor for a specific subset of ETS/AP-1-responsive genes, and as a modulator of the nuclear response to mitogen-activated protein kinase signaling cascades. It binds to DNA sequences containing the consensus nucleotide core sequence GGAA and involved in regulation of TNFRSF10B/DR5 expression through Ets-binding sequences on the TNFRSF10B/DR5 promoter

Sequence

Synthetic peptide located within the following region: SCIPFQEFDISGEHLCSMSLQEFTRAAGSAGQLLYSNLQHLKWNGQCSSD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service