Skip to Content
Merck
All Photos(1)

Key Documents

AV38669

Sigma-Aldrich

Anti-NR1H3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Nuclear receptor subfamily 1, group H, member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

50 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR1H3(10062)

Immunogen

Synthetic peptide directed towards the N terminal region of human NR1H3

Biochem/physiol Actions

NR1H3 belongs to the NR1 subfamily of nuclear receptor superfamily. The NR1 members regulate transcription that is required in lipid homeostasis and inflammation. NR1H3 is also known as LXRα is highly expressed in liver, kidney and intestine. LXRs act as cholesterol sensors and protect the cells from cholesterol overload.

Sequence

Synthetic peptide located within the following region: MSLWLGAPVPDIPPDSAVELWKPGAQDASSQAQGGSSCILREEARMPHSA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyan Zhao et al.
The Journal of endocrinology, 204(3), 233-240 (2009-10-20)
The liver X receptors (LXRs) are nuclear receptors that are activated by endogenous oxysterols, oxidized derivatives of cholesterol. There are two isoforms of LXR, LXRalpha (NR1H3) and LXRbeta (NR1H2). Both LXRalpha and LXRbeta regulate gene expression by binding to DNA
Peter A Edwards et al.
Journal of lipid research, 43(1), 2-12 (2002-01-17)
During the last three years there have been a plethora of publications on the liver X-activated receptors (LXRalpha, NR1H3, and LXRbeta, NR1H2), the farnesoid X-activated receptor (FXR, NR1H4), and the pregnane X receptor (PXR, NR1I2) and the role these nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service