Skip to Content
Merck
All Photos(3)

Key Documents

AV38128

Sigma-Aldrich

Anti-IRF1 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IRF-1, Anti-Interferon regulatory factor 1, Anti-MAR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€484.00

€484.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€484.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€484.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

sheep, rabbit, guinea pig, rat, dog, human, mouse, bovine, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IRF1(3659)

General description

Interferon regulatory factor 1 (IRF1) is a transcription factor and gene trans-activator that activates interferon-β and various other target gene expression to help regulate cell processes such as the immune response, apoptosis, and tumor suppression.

Specificity

Anti-IRF1 (AB2) polyclonal antibody reacts with chicken, pig, canine, bovine, human, mouse, and rat interferon regulatory factor 1 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human IRF1

Application

Anti-IRF1 (AB2) polyclonal antibody is used to tag interferon regulatory factor 1 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of interferon regulatory factor 1 in gene expression of proteins involve in the immune response, apoptosis, and tumor suppression.

Biochem/physiol Actions

IRF1 is interferon regulatory factor 1, a member of the interferon regulatory transcription factor (IRF) family. IRF1 serves as an activator of interferons alpha and beta transcription, and in mouse it has been shown to be required for double-stranded RNA induction of these genes. IRF1 also functions as a transcription activator of genes induced by interferons alpha, beta, and gamma. Further, IRF1 has been shown to play roles in regulating apoptosis and tumor-suppression.

Sequence

Synthetic peptide located within the following region: MPITRMRMRPWLEMQINSNQIPGLIWINKEEMIFQIPWKHAAKHGWDINK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service