Skip to Content
Merck
All Photos(2)

Key Documents

AV36525

Sigma-Aldrich

Anti-DHX30 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DDX30, Anti-DEAH (Asp-Glu-Ala-His) box polypeptide 30, Anti-FLJ11214, Anti-KIAA0890

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€374.00

€374.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€374.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DHX30(22907)

Immunogen

Synthetic peptide directed towards the N terminal region of human DHX30

Biochem/physiol Actions

DHX30 (DDX30) is a member of putative ATP-dependent RNA helicases called the DEAD box proteins. It is involved in regulation of RNA secondary structure during translation initiation, nuclear and mitochondrial splicing and ribosome and spliceosome assembly. DHX30 associates with mitochondrial DNA and has a role in retinal development.

Sequence

Synthetic peptide located within the following region: AESGMAPGGPGEGDGSLVNASRDLLKEFPQPKNLLNSVIGRALGISHAKD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiong Qin et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 29(3), 298-309 (2012-10-27)
NR2E3 is an orphan nuclear receptor expressed exclusively in photoreceptor cells of the retina. NR2E3-specific modulators may prolong photoreceptor survival in patients with dry age-related macular degeneration and other forms of retinal degeneration. To definitively establish NR2E3 as a photoreceptor
Yousong Wang et al.
The Journal of biological chemistry, 281(35), 25791-25802 (2006-07-11)
Mitochondrial DNA (mtDNA) is packaged into bacterial nucleoid-like structures, each containing several mtDNA molecules. The distribution of nucleoids during mitochondrial fission and fusion events and during cytokinesis is important to the segregation of mitochondrial genomes in heteroplasmic cells bearing a
Frances V Fuller-Pace
Nucleic acids research, 34(15), 4206-4215 (2006-08-29)
The DExD/H box family of proteins includes a large number of proteins that play important roles in RNA metabolism. Members of this family have been shown to act as RNA helicases or unwindases, using the energy from ATP hydrolysis to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service