Skip to Content
Merck
All Photos(2)

Documents

AV35242

Sigma-Aldrich

Anti-TRPM5 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-LTRPC5, Anti-MTR1, Anti-Transient receptor potential cation channel, subfamily M, member 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

131 kDa

species reactivity

mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPM5(29850)

General description

TRPM5 is a Ca+-activated channel that is involved in the transduction of umami, sweet and bitter tastes. Voltage, temperature, acidic pH and phosphoinositides are known to modulate TRPM5 function. It regulates mucin secretion in colon and pheromone transduction in olfactory epithelia. TRPM5 has been studied as a target for obesity treatment.
Rabbit Anti-TRPM5 antibody recognizes human, canine, and mouse TRPM5.

Immunogen

Synthetic peptide directed towards the N terminal region of human TRPM5

Application

Rabbit Anti-TRPM5 antibody is suitable for western blot applications at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

TRPM5 is a voltage-modulated Ca(2+)-activated, monovalent cation channel (VCAM) that mediates a transient membrane depolarization and plays a central role in taste transduction. It is activated by arachidonic acid in vitro. It may be involved in perception of bitter, sweet and umami tastes. It may also be involved in sensing semiochemicals.

Sequence

Synthetic peptide located within the following region: EKHISEQRAGYGGTGSIEIPVLCLLVNGDPNTLERISRAVEQAAPWLILV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Kyle Palmer et al.
Current topics in medicinal chemistry, 13(3), 247-257 (2013-02-26)
The disease of obesity is one of the greatest healthcare challenges of our time. The increasing urgency for effective treatment is driving an intensive search for new targets for anti-obesity drug discovery. The TRP channel super family represents a class
Fabián López et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(9), 3268-3278 (2014-02-28)
Growing evidence suggests that the main olfactory epithelium contains a subset of olfactory sensory neurons (OSNs) responding to pheromones. One candidate subpopulation expresses the calcium activated cation channel TRPM5 (transient receptor potential channel M5). Using GFP driven by the TRPM5
Yi-Hong Li et al.
eLife, 12 (2024-06-05)
Tuft cells are a group of rare epithelial cells that can detect pathogenic microbes and parasites. Many of these cells express signaling proteins initially found in taste buds. It is, however, not well understood how these taste signaling proteins contribute
E R Liman
Handbook of experimental pharmacology, (179)(179), 287-298 (2007-01-16)
TRPM5 is a cation channel that it is essential for transduction of bitter, sweet and umami tastes. Signaling of these tastes involves the activation of G protein-coupled receptors that stimulate phospholipase C (PLC) beta2, leading to the breakdown of phosphatidylinositol
Sandra Mitrovic et al.
eLife, 2, e00658-e00658 (2013-06-07)
Mucin 5AC (MUC5AC) is secreted by goblet cells of the respiratory tract and, surprisingly, also expressed de novo in mucus secreting cancer lines. siRNA-mediated knockdown of 7343 human gene products in a human colonic cancer goblet cell line (HT29-18N2) revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service