Skip to Content
Merck
All Photos(4)

Key Documents

AV34378

Sigma-Aldrich

Anti-FHL1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Four and a half LIM domains 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€382.00

€382.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€382.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€382.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

human, bovine, sheep, mouse, dog, guinea pig, horse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FHL1(2273)

General description

The previously assigned protein identifier Q6IB30 has been merged into Q13642. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the C terminal region of human FHL1

Biochem/physiol Actions

LIM proteins, named for ′LIN11, ISL1, and MEC3,′ are defined by the possession of a highly conserved double zinc finger motif called the LIM domain. FHL1 may play an important role during the early stages of skeletal muscle differentiation, specifically in alpha5beta1-integrin-mediated signaling pathways.

Sequence

Synthetic peptide located within the following region: YYCVDCYKNFVAKKCAGCKNPITGFGKGSSVVAYEGQSWHDYCFHCKKCS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed Gagaoua et al.
Meat science, 145, 308-319 (2018-07-18)
Reverse Phase Protein Arrays (RPPA) were applied for the quantification and validation of protein biomarkers of beef qualities on M. longissimus thoracis sampled early post-mortem from young Charolais bulls. pHu was related to six proteins, three of which are glycolytic
Mohammed Gagaoua et al.
Foods (Basel, Switzerland), 9(9) (2020-08-30)
This study evaluated the potential of a panel of 20 protein biomarkers, quantified by Reverse Phase Protein Array (RPPA), to explain and predict two important meat quality traits, these being beef tenderness assessed by Warner-Bratzler shear force (WBSF) and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service