Skip to Content
Merck
All Photos(1)

Documents

AV32371

Sigma-Aldrich

Anti-SKIL antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-SKI-like oncogene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SKIL(6498)

General description

SKIL forms a part of the SMAD pathway that regulates TGF-β mediated cell growth and differentiation. Rabbit Anti-SKIL antibody recognizes canine, human, mouse, rat, and bovine TGF-β.

Immunogen

Synthetic peptide directed towards the N terminal region of human SKIL

Application

Rabbit Anti-SKIL antibody can be used for western blot applications at a concentration of 2.5μg/ml.

Biochem/physiol Actions

SKIL belongs to the SKI family and may have regulatory role in cell division or differentiation in response to extracellular signals.

Sequence

Synthetic peptide located within the following region: IHVNGKTINKVPTVKKEHLDDYGEAPVETDGEHVKRTCTSVPETLHLNPS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service