Skip to Content
Merck
All Photos(2)

Key Documents

AV32341

Sigma-Aldrich

Anti-PSMD4 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Proteasome (prosome, macropain) 26S subunit, non-ATPase, 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

human, rat, horse, sheep, pig, rabbit, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSMD4(5710)

General description

PSDM4 is the non-ATPase subunit of the proteasomal 19S regulatory complex that is involved in proteasome-substrate identification. Porcine fertilization studies have reported that PSMD4 regulates sperm-zona pellucida (ZP) penetration. Furthermore, pharmacogenomic analyses have revealed that high expression of PSDM4 is linked to adverse clinical outcomes in myeloma patients undergoing bortezomib therapy.
Rabbit Anti-PSMD4 (AB1) antibody recognizes human, rat, rabbit, bovine, mouse, and pig PSMD4.

Immunogen

Synthetic peptide directed towards the C terminal region of human PSMD4

Application

Rabbit Anti-PSMD4 (AB1) antibody can be used for western blot assays at a concentration of 0.5μg/ml.

Biochem/physiol Actions

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. PSMD4 encodes one of the non-ATPase subunits of the 19S regulator lid.

Sequence

Synthetic peptide located within the following region: VMQDPEFLQSVLENLPGVDPNNEAIRNAMGSLASQATKDGKKDKKEEDKK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John D Shaughnessy et al.
Blood, 118(13), 3512-3524 (2011-06-02)
Gene expression profiling (GEP) of purified plasma cells 48 hours after thalidomide and dexamethasone test doses showed these agents' mechanisms of action and provided prognostic information for untreated myeloma patients on Total Therapy 2 (TT2). Bortezomib was added in Total
Young-Joo Yi et al.
Cell and tissue research, 341(2), 325-340 (2010-06-08)
Proteolysis of ubiquitinated sperm and oocyte proteins by the 26S proteasome is necessary for the success of mammalian fertilization, including but not limited to acrosomal exocytosis and sperm-zona pellucida (ZP) penetration. The present study examined the role of PSMD4, an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service