Skip to Content
Merck
All Photos(3)

Key Documents

AV32256

Sigma-Aldrich

Anti-NR2F6 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Nuclear receptor subfamily 2, group F, member 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€569.00

€569.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€569.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€569.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

43 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR2F6(2063)

General description

NR2F6 is a nuclear receptor that functions as PKC substrate. This receptor is known to regulate the responses of CD4+ T cell activation. Rabbit Anti-NR2F6 (AB1) antibody recognizes canine, human, mouse, and rat NR2F6.

Immunogen

Synthetic peptide directed towards the N terminal region of human NR2F6

Application

Rabbit Anti-NR2F6 (AB1) antibody can be used for western blot assays at a concentration of 0.1-5.0μg/ml.

Biochem/physiol Actions

NR2F6 is a nuclear orphan receptor that belongs to the COUP-TF subfamily

Sequence

Synthetic peptide located within the following region: MAMVTGGWGGPGGDTNGVDKAGGYPRAAEDDSASPPGAASDAEPGDEERP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natascha Hermann-Kleiter et al.
Immunity, 29(2), 205-216 (2008-08-15)
The protein kinase C (PKC) family of serine-threonine kinases plays a central role in T lymphocyte activation. Here, we identify NR2F6, a nuclear zinc-finger orphan receptor, as a critical PKC substrate and essential regulator of CD4(+) T cell activation responses.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service