Skip to Content
Merck
All Photos(1)

Key Documents

AV32186

Sigma-Aldrich

Anti-KLF3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Kruppel-like factor 3 (basic)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€287.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

rabbit, guinea pig, horse, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF3(51274)

General description

KLF3 is a transcriptional factor that regulates muscle-specific genes. It is known to interact with serum response factor (SRF) through KLF binding sites.
Rabbit Anti-KLF3 antibody recognizes canine, human, mouse, rat, pig, chicken, and bovine KLF3.

Immunogen

Synthetic peptide directed towards the N terminal region of human KLF3

Application

Rabbit Anti-KLF3 antibody can be used for western blot applications at a concentration of 1.25μg/ml.

Biochem/physiol Actions

KLF3 is a zinc finger transcription factor that is known to function as a potent transcriptional repressor

Sequence

Synthetic peptide located within the following region: LSHGIQMEPVDLTVNKRSSPPSAGNSPSSLKFPSSHRRASPGLSMPSSSP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnes Fütterer et al.
Cell death & disease, 12(7), 637-637 (2021-06-23)
Embryonic stem cell (ESC) differentiation and somatic cell reprogramming are biological processes governed by antagonistic expression or repression of a largely common set of genes. Accurate regulation of gene expression is thus essential for both processes, and alterations in RNA
Charis L Himeda et al.
Molecular and cellular biology, 30(14), 3430-3443 (2010-04-21)
This study identifies KLF3 as a transcriptional regulator of muscle genes and reveals a novel synergistic interaction between KLF3 and serum response factor (SRF). Using quantitative proteomics, KLF3 was identified as one of several candidate factors that recognize the MPEX

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service