Skip to Content
Merck
All Photos(3)

Key Documents

AV31444

Sigma-Aldrich

Anti-HOXB5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HHO.C10, Anti-HOX2, Anti-HOX2A, Anti-HU-1, Anti-Homeobox B5, Anti-Hox2.1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

30 kDa

species reactivity

horse, mouse, bovine, rat, dog, guinea pig, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HOXB5(3215)

General description

HOXB5 is known to regulate the development of gut neural crest cells in human embryos[1]. It is also known to function as a transcriptional switch for vascular endothelial cell differentiation[2].
Rabbit Anti-HOXB5 antibody binds to canine, human, mouse, and rat HOXB5.

Immunogen

Synthetic peptide directed towards the N terminal region of human HOXB5

Application

Rabbit Anti-HOXB5 antibody can be used for western blot applications at 0.5μg/ml.

Biochem/physiol Actions

HOXB5 belongs to the homeobox family. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXB genes located in a cluster on chromosome 17. The exact role of this gene has yet to be determined.This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in lung and gut development. Increased expression of this gene is associated with a distinct biologic subset of acute myeloid leukemia (AML) and the occurrence of bronchopulmonary sequestration (BPS) and congenital cystic adenomatoid malformation (CCAM) tissue.

Sequence

Synthetic peptide located within the following region: MSSYFVNSFSGRYPNGPDYQLLNYGSGSSLSGSYRDPAAMHTGSYGYNYN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Fu et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 228(1), 1-10 (2003-09-02)
HOX genes from paralogous groups 4 and 5 are particularly relevant to the gut neuromusculature development because these genes are expressed at the splanchnic mesoderm surrounding the gut diverticulum, and at the level of the neural tube from where the
Yaxu Wu et al.
Molecular and cellular biology, 23(16), 5680-5691 (2003-08-05)
Endothelial cells differentiate from mesoderm-derived precursors to initiate the earliest events in vascular development. Although the signaling events that regulate the successive steps of vascular development are known in some detail, the transcriptional processes that regulate the first steps in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service