Skip to Content
Merck
All Photos(1)

Key Documents

AV100917

Sigma-Aldrich

Anti-CSDA (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Cold shock domain protein A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€287.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

human, pig, horse, rabbit, bovine, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSDA(8531)

Immunogen

Synthetic peptide directed towards the C terminal region of human CSDA

Application

Anti-CSDA (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

CSDA is a member of RNA-and DNA-binding cold-shock-domain (CSD) family that regulates the concentration of EGR1 to mediate luteinizing hormone β subunit transcription. It regulates Bcr-Abl effector, and also regulates cell proliferation and transformation in chronic myeloid leukemia.

Sequence

Synthetic peptide located within the following region: GPPRPRPAPAVGEAEDKENQQATSGPNQPSVRRGYRRPYNYRRRPRPPNA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Sears et al.
Cell death & disease, 1, e93-e93 (2010-01-01)
One proposed strategy to suppress the proliferation of imatinib-resistant cells in chronic myeloid leukemia (CML) is to inhibit key proteins downstream of Bcr-Abl. The PI3K/Akt pathway is activated by Bcr-Abl and is specifically required for the growth of CML cells.
Wenming Pan et al.
Cell death and differentiation, 26(2), 291-305 (2018-05-18)
Hepatic ischemia/reperfusion injury (IRI) is a common cause of morbidity and mortality in liver transplantation settings and involves severe cell death and inflammatory responses. MicroRNA-191 has recently been reported to be abnormally expressed in hepatocellular carcinoma and other liver diseases
Theodore R Chauvin et al.
Biology of reproduction, 86(2), 53-53 (2011-11-05)
Gonadotropin-releasing hormone (GnRH), a hypothalamic neurohormone, regulates transcription of Lhb in gonadotrophs indirectly through transient induction and accumulation of EGR1, a zinc finger transcription factor. AlphaT3 and LbetaT2 cell lines model gonadotrophs at two distinct stages of development, prenatal and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service