Skip to Content
Merck
All Photos(2)

Documents

AV100775

Sigma-Aldrich

Anti-TCF4 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Transcription factor 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

71 kDa

species reactivity

mouse, horse, guinea pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TCF4(6925)

General description

TCF-4 belongs to the Tcf family of transcription factors that activate genes induced by the Wnt pathway.

Immunogen

Synthetic peptide directed towards the N terminal region of human TCF4

Application

Anti-TCF4 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

TCF-4 mediates the transformation of epithelial cells of colon in the absence of APC gene expression. The expression of TCF-4 is required to establish the proliferative progenitors to form the crypts of embryonic small intestine. TCF-4 collaborates with b-catenin to regulate the colorectal transformation process. Lack of TCF-4 expression results in developmental delay and intellectual disability termed as Pitt-Hopkins syndrome.

Sequence

Synthetic peptide located within the following region: MHHQQRMAALGTDKELSDLLDFSAMFSPPVSSGKNGPTSLASGHFTGSNV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc van de Wetering et al.
Cell, 111(2), 241-250 (2002-11-01)
The transactivation of TCF target genes induced by Wnt pathway mutations constitutes the primary transforming event in colorectal cancer (CRC). We show that disruption of beta-catenin/TCF-4 activity in CRC cells induces a rapid G1 arrest and blocks a genetic program
J David Sweatt
Experimental & molecular medicine, 45, e21-e21 (2013-05-04)
TCF4 (transcription factor 4; E2-2, ITF2) is a transcription factor that when haplo-insufficient causes Pitt-Hopkins Syndrome (PTHS), an autism-spectrum disorder that is associated with pervasive developmental delay and severe intellectual disability. The TCF4 gene is also a risk factor with
V Korinek et al.
Nature genetics, 19(4), 379-383 (1998-08-11)
Mutations of the genes encoding APC or beta-catenin in colon carcinoma induce the constitutive formation of nuclear beta-catenin/Tcf-4 complexes, resulting in activated transcription of Tcf target genes. To study the physiological role of Tcf-4 (which is encoded by the Tcf7/2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service