Skip to Content
Merck
All Photos(3)

Key Documents

AV09002

Sigma-Aldrich

Anti-SerPINH1 antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€287.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINH1(871)

Immunogen

Synthetic peptide directed towards the C terminal region of human SERPINH1

Application

Anti- SerPINH1 antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

SerPINH1 (Hsp47) is a molecular chaperone that regulates protein folding of type I and type IV procollagen in the endoplasmic reticulum. The activity of Hsp47 is critical for the development of well-organized cartilage and formation of normal endochondral bone.

Sequence

Synthetic peptide located within the following region: DIYGREELRSPKLFYADHPFIFLVRDTQSGSLLFIGRLVRLKGDKMRDEL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Widmer et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(33), 13243-13247 (2012-08-01)
Collagen is the most abundant protein in animals and is a major component of the extracellular matrix in tissues such as skin and bone. A distinctive structural feature of all collagen types is a unique triple-helical structure formed by tandem
Yusaku Masago et al.
Journal of cell science, 125(Pt 5), 1118-1128 (2012-04-12)
Heat shock protein 47 kDa (Hsp47) is considered as a molecular chaperone essential for the correct folding of type I and type IV procollagen in the ER. However, the function of Hsp47 for other types of procollagen and its importance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service