Skip to Content
Merck
All Photos(1)

Key Documents

AC-0369A

Sigma-Aldrich

HMGA2 (EP398) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 ML
€303.00

€303.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.1 ML
€303.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€303.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:60

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

High-mobility group AT-hook 2 (HMGA2) is a chromatin factor that regulates transcription and cellular differentiation during embryonic development. HMGA2 helps to regulate the cell cycle and plays an important role in chromosome condensation during the meiotic G2/M transition of spermatocytes. It also interacts with transcription factor E4F1, protein kinase NEK2, and the promoter of the nucleotide excision-repair gene ERCC1 to negatively modulate its activity. HMGA2 expression in adult tissues is often associated with benign and malignant tumor formation. HMGA2 is expressed in most conventional and intramuscular lipomas, 84 and 100%, respectively and can aid in differentiating between lipomas from dedifferentiated liposarcomas and distinguishing areas of tumor from normal adipose tissue. In mesenchymal tumors, HMGA2 is expressed in 88% of benign fibrous histiocytoma, 90% of nodular fasciitis, and 90% of vulvovaginal angiomyxoma. Increased expression of HMGA2 occurs in other human malignancies, including pancreatic, gastric, oral squamous cell, ovarian, and a subset of breast cancers. In thyroid carcinomas, upregulation of HMGA2 can distinguish between benign and malignant follicular neoplasias. Further, HMGA2 overexpression is often found in non-small cell lung cancer and could be used as a marker for lung carcinomas. The presence of HMGA2 in cancer is correlative with metastasis and poor prognosis.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and < 0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service