Skip to Content
Merck
All Photos(1)

Key Documents

AC-0363

Sigma-Aldrich

SPINK1 (TATI, PSTI) (EP401) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€903.00

€903.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€903.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€903.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Serine protease inhibitor Kazal-type 1 (SPINK1), also known as tumor-associated trypsin inhibitor (TATI) and pancreatic secretory trypsin inhibitor (PSTI) is a cytosolic trypsin inhibitor secreted by pancreatic acinar cells and foveolar cells of the stomach that protects the gastrointestinal system from premature trypsinogen auto-digestion. Mutations in SPINK1 are associated with hereditary pancreatitis. Recently, SPINK1 demonstrated utility in differentiating hepatocellular carcinoma (HCC) from precancerous lesions. SPINK1 is detected in all hereditary haemochromatosis HCC and most (79%) of HCC cases, but limited to the luminal surface of large bile ducts in normal liver. SPINK1 expression is also detected in neoplasms of the ovary, bladder, kidney, pancreas, colon, lung, and breast. High expression of SPINK1 is often associated with advanced tumor staging, making it a useful prognostic marker.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and < 0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service