Skip to Content
Merck
All Photos(1)

Key Documents

AC-0362

Sigma-Aldrich

ACTH (EP390) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€903.00

€903.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€903.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€903.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Adrenocorticotropic hormone (ACTH), also known as corticotropin, is produced from corticotropes in the anterior lobe of the pituitary gland. ACTH is released by corticotropin-releasing hormone stimulation by the hypothalamus in response to biological stress. Identifying ACTH is useful in the classification of pituitary tumors and diseases. Cushing′s disease derived from ACTH producing adenomas represents 10-15% of pituitary tumors. By immunohistochemistry (IHC), these tumors are immunoreactive for ACTH, beta-Lipotropin, and beta-endorphin. ACTH is also detected in a subset of growth hormone (GH), thyroid-stimulating hormone (TSH) and follicle-stimulating hormone (FSH) secreting adenomas. With the exception for ACTH secreting adenomas, most functioning adenomas are usually macroadenomas; tumors larger than 10 mm. A combination of ACTH IHC and reticulin staining also provides utility in the differential diagnosis between pituitary adenoma versus corticotroph hyperplasia. Breakdown of acinar structure and ACTH immunoreactivity are suggestive of adenoma.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &lt0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service