Skip to Content
Merck
All Photos(1)

Key Documents

ABK0018

Sigma-Aldrich

AfliBridge ELISA

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€1,130.00

€1,130.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€1,130.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

€1,130.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

purified antibody

Quality Level

species reactivity

mouse, human

shipped in

wet ice

storage temp.

2-8°C

General description

A Protein Conformational Array (PCA) in a sandwich ELISA format where the plate is coated with a panel of antibodies raised against peptides derived from the full-length protein sequence of Aflibercept. Taken individually, each of these antibodies is strongly antigenic to the peptide sequence that was used in its production. However, when these peptides are incorporated into a full length correctly folded protein, the antigenicity of many of them is masked by the three-dimensional structure of the protein and only a limited number of the antibodies respond. The result is a histogram which can be likened to a ‘fingerprint′ for correctly folded Aflibercept. For a Eylea Biosimilar, if the protein is correctly folded and glycosylated, the ‘fingerprint′ will match that of Aflibercept. If it is not correctly folded, previously masked peptide sequences will be exposed and will be recognized by the antibody made to that exposed sequence. In this way, changes in the ‘fingerprint′ generated by the ELISA will point out differences between the Biosimilar and authentic Aflibercept.

Application

For research use only. Not for use in diagnostic procedures.

Other Notes

ELISA kit contains 3 96-well microplates and all the components necessary to complete an ELISA test.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service